Assay Details
| Gene Symbol: Polr2g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004874400.2 | XP_004874457.1 | 7 - 8 | 584 | 54 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Polr2g| Gene Symbol: Polr2g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004874400.2 | XP_004874457.1 | 7 - 8 | 584 | 54 | |
Set Membership: |
|