Assay Details
| Gene Symbol: Ric1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021253021.1 | XP_021108680.1 | 23 - 24 | 3754 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ric1| Gene Symbol: Ric1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021253021.1 | XP_021108680.1 | 23 - 24 | 3754 | 64 | |
Set Membership: |
|