Assay Details
| Gene Symbol: Timm22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004857098.3 | XP_004857155.1 | 2 - 3 | 521 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Timm22| Gene Symbol: Timm22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004857098.3 | XP_004857155.1 | 2 - 3 | 521 | 63 | |
Set Membership: |
|