Assay Details
| Gene Symbol: Lrrc17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004839730.2 | XP_004839787.1 | 3 - 4 | 1216 | 72 | |
| XM_021253959.1 | XP_021109618.1 | 4 - 5 | 1286 | 72 | ||
More Information
Set Membership: |
|

Lrrc17
Genomic Map