Assay Details
| Gene Symbol: Cdk7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872832.2 | XP_004872889.1 | 8 - 9 | 1266 | 69 | |
| XM_004872833.2 | XP_004872890.1 | 8 - 9 | 1255 | 69 | ||
| XM_021250691.1 | XP_021106350.1 | 8 - 9 | 1274 | 69 | ||
More Information
Set Membership: |
|

Cdk7
Genomic Map