Assay Details
| Gene Symbol: Afmid | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004860818.3 | XP_004860875.1 | 2 - 3 | 259 | 64 | |
| XM_004860820.3 | XP_004860877.1 | 2 - 3 | 259 | 64 | ||
| XM_004860821.3 | XP_004860878.1 | 2 - 3 | 259 | 64 | ||
| XM_021241042.1 | XP_021096701.1 | 2 - 3 | 271 | 64 | ||
More Information
Set Membership: |
|

Afmid
Genomic Map