Assay Details
| Gene Symbol: Tram1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842062.3 | XP_004842119.1 | 7 - 8 | 801 | 67 | |
| XM_021256028.1 | XP_021111687.1 | 7 - 8 | 798 | 67 | ||
More Information
Set Membership: |
|

Tram1
Genomic Map