Assay Details
| Gene Symbol: Blvrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004874205.3 | XP_004874262.1 | 2 - 3 | 440 | 54 | |
| XM_021251693.1 | XP_021107352.1 | 2 - 3 | 470 | 54 | ||
| XM_021251694.1 | XP_021107353.1 | 2 - 3 | 467 | 54 | ||
More Information
Set Membership: |
|

Blvrb
Genomic Map