Assay Details
| Gene Symbol: Asb15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004856472.2 | XP_004856529.2 | 7 - 8 | 841 | 77 | |
| XM_021237631.1 | XP_021093290.1 | 7 - 8 | 841 | 77 | ||
More Information
Set Membership: |
|

Asb15
Genomic Map