Assay Details
| Gene Symbol: Plvap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004873226.3 | XP_004873283.1 | 2 - 3 | 514 | 66 | |
| XM_004873227.2 | XP_004873284.1 | 2 - 3 | 514 | 66 | ||
More Information
Set Membership: |
|

Plvap
Genomic Map