Assay Details
| Gene Symbol: Nnat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867797.3 | XP_004867854.1 | 1 - 2 | 272 | 75 | |
| XM_013066737.2 | XP_012922191.1 | 1 - 2 | 274 | 75 | ||
More Information
Set Membership: |
|

Nnat
Genomic Map