Assay Details
| Gene Symbol: Timp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004875119.3 | XP_004875176.1 | 1 - 2 | 439 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Timp4| Gene Symbol: Timp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004875119.3 | XP_004875176.1 | 1 - 2 | 439 | 70 | |
Set Membership: |
|