Assay Details
| Gene Symbol: Znf827 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004868913.3 | XP_004868970.1 | 10 - 11 | 3186 | 66 | |
| XM_004868916.2 | XP_004868973.1 | 10 - 11 | 3186 | 66 | ||
| XM_004868919.3 | XP_004868976.1 | 10 - 11 | 3186 | 66 | ||
| XM_013067124.2 | XP_012922578.1 | 10 - 11 | 3186 | 66 | ||
| XM_013067125.2 | XP_012922579.1 | 10 - 11 | 3186 | 66 | ||
| XM_013067126.2 | XP_012922580.1 | 10 - 11 | 3186 | 66 | ||
| XM_021247679.1 | XP_021103338.1 | 10 - 11 | 3186 | 66 | ||
More Information
Set Membership: |
|

Znf827
Genomic Map