Assay Details
| Gene Symbol: Pus7l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004863163.3 | XP_004863220.1 | 5 - 6 | 1451 | 80 | |
| XM_021243143.1 | XP_021098802.1 | 4 - 5 | 523 | 80 | ||
| XM_021243144.1 | XP_021098803.1 | 4 - 5 | 1147 | 80 | ||
More Information
Set Membership: |
|

Pus7l
Genomic Map