Assay Details
| Gene Symbol: Zdhhc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835053.3 | XP_004835110.1 | 4 - 5 | 727 | 64 | |
| XM_004835054.2 | XP_004835111.1 | 4 - 5 | 727 | 64 | ||
| XM_021257280.1 | XP_021112939.1 | 4 - 5 | 731 | 64 | ||
| XM_021257284.1 | XP_021112943.1 | 4 - 5 | 727 | 64 | ||
| XM_021257290.1 | XP_021112949.1 | 4 - 5 | 727 | 64 | ||
| XM_021257293.1 | XP_021112952.1 | 4 - 5 | 727 | 64 | ||
| XM_021257297.1 | XP_021112956.1 | 4 - 5 | 727 | 64 | ||
| XM_021257303.1 | XP_021112962.1 | 4 - 5 | 727 | 64 | ||
| XM_021257311.1 | XP_021112970.1 | 4 - 5 | 731 | 64 | ||
| XM_021257318.1 | XP_021112977.1 | 4 - 5 | 731 | 64 | ||
More Information
Set Membership: |
|

Zdhhc3
Genomic Map