Assay Details
| Gene Symbol: Zranb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004838751.3 | XP_004838808.1 | 5 - 6 | 1694 | 82 | |
| XM_021253299.1 | XP_021108958.1 | 5 - 6 | 688 | 82 | ||
More Information
Set Membership: |
|

Zranb1
Genomic Map