Assay Details
| Gene Symbol: Tpmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847624.3 | XP_004847681.1 | 6 - 7 | 1224 | 85 | |
| XM_013073267.2 | XP_012928721.1 | 6 - 7 | 1222 | 85 | ||
| XM_021260586.1 | XP_021116245.1 | 5 - 6 | 592 | 85 | ||
| XM_021260587.1 | XP_021116246.1 | 6 - 7 | 1014 | 85 | ||
| XM_021260588.1 | XP_021116247.1 | 6 - 7 | 845 | 85 | ||
| XM_021260589.1 | XP_021116248.1 | 5 - 6 | 942 | 85 | ||
| XM_021260590.1 | XP_021116249.1 | 6 - 7 | 1223 | 85 | ||
| XM_021260591.1 | XP_021116250.1 | 4 - 5 | 442 | 85 | ||
More Information
Set Membership: |
|

Tpmt
Genomic Map