Assay Details
| Gene Symbol: Tada2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847331.2 | XP_004847388.1 | 1 - 2 | 314 | 100 | |
| XM_021260319.1 | XP_021115978.1 | 1 - 2 | 314 | 100 | ||
More Information
Set Membership: |
|

Tada2b
Genomic Map