Assay Details
| Gene Symbol: Map9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004868555.3 | XP_004868612.1 | 8 - 9 | 1280 | 72 | |
| XM_004868556.3 | XP_004868613.2 | 8 - 9 | 1277 | 72 | ||
| XM_021247448.1 | XP_021103107.1 | 8 - 9 | 1280 | 72 | ||
| XM_021247449.1 | XP_021103108.1 | 6 - 7 | 1075 | 72 | ||
| XM_021247450.1 | XP_021103109.1 | 6 - 7 | 965 | 72 | ||
More Information
Set Membership: |
|

Map9
Genomic Map