Assay Details
| Gene Symbol: Styk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845892.2 | XP_004845949.1 | 8 - 9 | 1721 | 62 | |
| XM_021259181.1 | XP_021114840.1 | 8 - 9 | 1741 | 62 | ||
| XM_021259182.1 | XP_021114841.1 | 8 - 9 | 1464 | 62 | ||
| XM_021259183.1 | XP_021114842.1 | 8 - 9 | 1410 | 62 | ||
| XM_021259184.1 | XP_021114843.1 | 8 - 9 | 2116 | 62 | ||
| XM_021259185.1 | XP_021114844.1 | 8 - 9 | 2250 | 62 | ||
| XM_021259186.1 | XP_021114845.1 | 8 - 9 | 2047 | 62 | ||
| XM_021259187.1 | XP_021114846.1 | 7 - 8 | 1954 | 62 | ||
| XM_021259188.1 | XP_021114847.1 | 7 - 8 | 3011 | 62 | ||
| XM_021259189.1 | XP_021114848.1 | 7 - 8 | 1951 | 62 | ||
More Information
Set Membership: |
|

Styk1
Genomic Map