Assay Details
| Gene Symbol: Sucnr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004834468.3 | XP_004834525.2 | 5 - 6 | 506 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sucnr1| Gene Symbol: Sucnr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004834468.3 | XP_004834525.2 | 5 - 6 | 506 | 101 | |
Set Membership: |
|