Assay Details
| Gene Symbol: Rab39b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004875173.3 | XP_004875230.1 | 3 - 4 | 1297 | 88 | |
| XM_021252442.1 | XP_021108101.1 | 3 - 4 | 790 | 88 | ||
More Information
Set Membership: |
|

Rab39b
Genomic Map