Assay Details
| Gene Symbol: Slc22a18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004852114.3 | XP_004852171.1 | 4 - 5 | 571 | 62 | |
| XM_004852115.3 | XP_004852172.1 | 4 - 5 | 553 | 62 | ||
| XM_004852116.3 | XP_004852173.1 | 4 - 5 | 544 | 62 | ||
| XM_004852117.3 | XP_004852174.1 | 4 - 5 | 521 | 62 | ||
| XM_004852118.3 | XP_004852175.1 | 4 - 5 | 522 | 62 | ||
| XM_013075002.2 | XP_012930456.1 | 4 - 5 | 571 | 62 | ||
| XM_013075003.2 | XP_012930457.1 | 4 - 5 | 571 | 62 | ||
| XM_021264437.1 | XP_021120096.1 | 3 - 4 | 420 | 62 | ||
| XM_021264438.1 | XP_021120097.1 | 4 - 5 | 572 | 62 | ||
More Information
Set Membership: |
|

Slc22a18
Genomic Map