Assay Details
| Gene Symbol: Rnaseh2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004852382.2 | XP_004852439.1 | 2 - 3 | 402 | 75 | |
| XM_021264579.1 | XP_021120238.1 | 2 - 3 | 407 | 75 | ||
More Information
Set Membership: |
|

Rnaseh2c
Genomic Map