Assay Details
| Gene Symbol: Serinc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004860151.2 | XP_004860208.1 | 4 - 5 | 572 | 59 | |
| XM_004860152.2 | XP_004860209.1 | 4 - 5 | 572 | 59 | ||
More Information
Set Membership: |
|

Serinc1
Genomic Map