Assay Details
Gene Symbol: LOC101698131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013074067.2 | XP_012929521.2 | 2 - 3 | 205 | 77 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013074067.2 transcripts.
Set Membership: |
![]() ![]() |
