Assay Details
Gene Symbol: LOC101698131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013074067.2 | XP_012929521.2 | 4 - 5 | 438 | 71 | |
XM_021260287.1 | XP_021115946.1 | 4 - 5 | 549 | 71 | ||
XM_021260293.1 | XP_021115952.1 | 4 - 5 | 549 | 71 | ||
XM_021260299.1 | XP_021115958.1 | 4 - 5 | 549 | 71 | ||
XM_021260305.1 | XP_021115964.1 | 4 - 5 | 549 | 71 | ||
XM_021260307.1 | XP_021115966.1 | 4 - 5 | 549 | 71 | ||
XM_021260309.1 | XP_021115968.1 | 3 - 4 | 325 | 71 | ||
XM_021260311.1 | XP_021115970.1 | 4 - 5 | 549 | 71 | ||
XM_021260320.1 | XP_021115979.1 | 4 - 5 | 549 | 71 | ||
XM_021260324.1 | XP_021115983.1 | 4 - 5 | 549 | 71 | ||
XM_021260333.1 | XP_021115992.1 | 4 - 5 | 549 | 71 | ||
XM_021260341.1 | XP_021116000.1 | 4 - 5 | 549 | 71 | ||
XM_021260348.1 | XP_021116007.1 | 4 - 5 | 549 | 71 | ||
XM_021260358.1 | XP_021116017.1 | 4 - 5 | 549 | 71 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021260309.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
