Assay Details
| Gene Symbol: Rnf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004846659.2 | XP_004846716.1 | 4 - 5 | 1212 | 77 | |
| XM_013072968.2 | XP_012928422.1 | 4 - 5 | 577 | 77 | ||
| XM_013072969.2 | XP_012928423.1 | 3 - 4 | 471 | 77 | ||
More Information
Set Membership: |
|

Rnf8
Genomic Map