Assay Details
Gene Symbol: Ube2g2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004842506.3 | XP_004842563.1 | 4 - 5 | 327 | 68 | |
XM_013071378.2 | XP_012926832.1 | 2 - 3 | 362 | 68 | ||
XM_021256392.1 | XP_021112051.1 | 4 - 5 | 353 | 68 | ||
XM_021256393.1 | XP_021112052.1 | 5 - 6 | 370 | 68 | ||
XM_021256394.1 | XP_021112053.1 | 5 - 6 | 399 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
