Assay Details
| Gene Symbol: Pwwp2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004838703.2 | XP_004838760.1 | 1 - 2 | 164 | 96 | |
| XM_004838707.2 | XP_004838764.1 | 1 - 2 | 164 | 96 | ||
More Information
Set Membership: |
|

Pwwp2b
Genomic Map