Assay Details
| Gene Symbol: Pwwp2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004838703.2 | XP_004838760.1 | 2 - 3 | 1846 | 80 | |
| XM_021253229.1 | XP_021108888.1 | 2 - 3 | 2986 | 80 | ||
| XM_021253230.1 | XP_021108889.1 | 2 - 3 | 3051 | 80 | ||
| XM_021253231.1 | XP_021108890.1 | 2 - 3 | 2867 | 80 | ||
More Information
Set Membership: |
|

Pwwp2b
Genomic Map