Assay Details
| Gene Symbol: Lrrc57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004861659.2 | XP_004861716.1 | 2 - 3 | 370 | 69 | |
| XM_021241770.1 | XP_021097429.1 | 2 - 3 | 289 | 69 | ||
| XM_021241771.1 | XP_021097430.1 | 2 - 3 | 364 | 69 | ||
More Information
Set Membership: |
|

Lrrc57
Genomic Map