Assay Details
| Gene Symbol: Aagab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855596.2 | XP_004855653.1 | 5 - 6 | 611 | 70 | |
| XM_021237021.1 | XP_021092680.1 | 5 - 6 | 611 | 70 | ||
| XM_021237022.1 | XP_021092681.1 | 5 - 6 | 611 | 70 | ||
| XM_021237023.1 | XP_021092682.1 | 5 - 6 | 651 | 70 | ||
More Information
Set Membership: |
|

Aagab
Genomic Map