Assay Details
| Gene Symbol: Trim8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004866450.3 | XP_004866507.1 | 2 - 3 | 783 | 71 | |
| XM_021245559.1 | XP_021101218.1 | 2 - 3 | 926 | 71 | ||
More Information
Set Membership: |
|

Trim8
Genomic Map