Assay Details
| Gene Symbol: Tmem45a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004857978.3 | XP_004858035.1 | 4 - 5 | 926 | 82 | |
| XM_013077074.2 | XP_012932528.1 | 4 - 5 | 839 | 82 | ||
More Information
Set Membership: |
|

Tmem45a
Genomic Map