Assay Details
| Gene Symbol: Ccr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013076683.2 | XP_012932137.2 | 2 - 3 | 231 | 92 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013076683.2 transcripts.
Set Membership: |
|

Ccr6
Genomic Map