Assay Details
| Gene Symbol: Lcp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004844204.3 | XP_004844261.1 | 10 - 11 | 1170 | 63 | |
| XM_021257865.1 | XP_021113524.1 | 11 - 12 | 1187 | 63 | ||
| XM_021257866.1 | XP_021113525.1 | 9 - 10 | 1471 | 63 | ||
| XM_021257867.1 | XP_021113526.1 | 12 - 13 | 1298 | 63 | ||
| XM_021257868.1 | XP_021113527.1 | 9 - 10 | 1016 | 63 | ||
More Information
Set Membership: |
|

Lcp1
Genomic Map