Assay Details
| Gene Symbol: Elac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855167.2 | XP_004855224.1 | 2 - 3 | 306 | 84 | |
| XM_021236630.1 | XP_021092289.1 | 2 - 3 | 686 | 84 | ||
| XM_021236631.1 | XP_021092290.1 | 2 - 3 | 307 | 84 | ||
More Information
Set Membership: |
|

Elac1
Genomic Map