Assay Details
| Gene Symbol: Psma2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840246.3 | XP_004840303.1 | 2 - 3 | 264 | 61 | |
| XM_021254539.1 | XP_021110198.1 | 2 - 3 | 109 | 61 | ||
More Information
Set Membership: |
|

Psma2
Genomic Map