Assay Details
| Gene Symbol: Ubfd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855945.3 | XP_004856002.1 | 2 - 3 | 542 | 75 | |
| XM_004855946.3 | XP_004856003.1 | 2 - 3 | 534 | 75 | ||
| XM_021237322.1 | XP_021092981.1 | 2 - 3 | 543 | 75 | ||
| XM_021237323.1 | XP_021092982.1 | 2 - 3 | 540 | 75 | ||
More Information
Set Membership: |
|

Ubfd1
Genomic Map