Assay Details
| Gene Symbol: Zdhhc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842727.3 | XP_004842784.1 | 7 - 8 | 1021 | 66 | |
| XM_013071457.2 | XP_012926911.1 | 7 - 8 | 1062 | 66 | ||
| XM_013071458.2 | XP_012926912.1 | 7 - 8 | 887 | 66 | ||
| XM_021256616.1 | XP_021112275.1 | 7 - 8 | 869 | 66 | ||
More Information
Set Membership: |
|

Zdhhc7
Genomic Map