Assay Details
| Gene Symbol: B4galt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004871901.3 | XP_004871958.1 | 6 - 7 | 1361 | 59 | |
| XM_004871902.3 | XP_004871959.1 | 5 - 6 | 1162 | 59 | ||
| XM_004871903.3 | XP_004871960.1 | 5 - 6 | 1056 | 59 | ||
| XM_021249950.1 | XP_021105609.1 | 6 - 7 | 1364 | 59 | ||
More Information
Set Membership: |
|

B4galt2
Genomic Map