Assay Details
| Gene Symbol: P2ry2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021243278.1 | XP_021098937.1 | 1 - 2 | 348 | 88 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021243278.1 transcripts.
Set Membership: |
|

P2ry2
Genomic Map