Assay Details
| Gene Symbol: Limd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004868612.3 | XP_004868669.1 | 4 - 5 | 403 | 94 | |
| XM_021247489.1 | XP_021103148.1 | 4 - 5 | 406 | 94 | ||
| XM_021247490.1 | XP_021103149.1 | 4 - 5 | 338 | 94 | ||
| XM_021247491.1 | XP_021103150.1 | 4 - 5 | 320 | 94 | ||
| XM_021247492.1 | XP_021103151.1 | 4 - 5 | 435 | 94 | ||
| XM_021247493.1 | XP_021103152.1 | 4 - 5 | 468 | 94 | ||
| XM_021247494.1 | XP_021103153.1 | 4 - 5 | 300 | 94 | ||
| XM_021247495.1 | XP_021103154.1 | 4 - 5 | 404 | 94 | ||
More Information
Set Membership: |
|

Limd2
Genomic Map