Assay Details
| Gene Symbol: Fam19a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845370.3 | XP_004845427.1 | 2 - 3 | 454 | 71 | |
| XM_004845372.3 | XP_004845429.1 | 2 - 3 | 447 | 71 | ||
| XM_021258778.1 | XP_021114437.1 | 2 - 3 | 453 | 71 | ||
More Information
Set Membership: |
|

Fam19a5
Genomic Map