Assay Details
| Gene Symbol: Emp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867023.3 | XP_004867080.1 | 3 - 4 | 288 | 67 | |
| XM_004867024.3 | XP_004867081.1 | 3 - 4 | 361 | 67 | ||
| XM_021246263.1 | XP_021101922.1 | 3 - 4 | 789 | 67 | ||
More Information
Set Membership: |
|

Emp3
Genomic Map