Assay Details
| Gene Symbol: Coro1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004852235.3 | XP_004852292.1 | 10 - 11 | 1442 | 54 | |
| XM_013074952.2 | XP_012930406.1 | 11 - 12 | 1535 | 54 | ||
| XM_021264503.1 | XP_021120162.1 | 10 - 11 | 1296 | 54 | ||
| XM_021264504.1 | XP_021120163.1 | 10 - 11 | 1215 | 54 | ||
More Information
Set Membership: |
|

Coro1b
Genomic Map