Assay Details
| Gene Symbol: Plpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004848515.3 | XP_004848572.1 | 4 - 5 | 974 | 71 | |
| XM_004848516.3 | XP_004848573.1 | 4 - 5 | 983 | 71 | ||
| XM_021261419.1 | XP_021117078.1 | 4 - 5 | 572 | 71 | ||
More Information
Set Membership: |
|

Plpp1
Genomic Map