Assay Details
| Gene Symbol: Znf597 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004864744.3 | XP_004864801.1 | 4 - 5 | 1453 | 116 | |
| XM_004864745.3 | XP_004864802.1 | 4 - 5 | 1456 | 116 | ||
| XM_004864746.2 | XP_004864803.1 | 4 - 5 | 1511 | 116 | ||
| XM_021244395.1 | XP_021100054.1 | 4 - 5 | 1503 | 116 | ||
More Information
Set Membership: |
|

Znf597
Genomic Map