Assay Details
| Gene Symbol: Tinagl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004850815.2 | XP_004850872.1 | 3 - 4 | 468 | 63 | |
| XM_004850816.2 | XP_004850873.1 | 3 - 4 | 517 | 63 | ||
| XM_021263238.1 | XP_021118897.1 | 3 - 4 | 468 | 63 | ||
More Information
Set Membership: |
|

Tinagl1
Genomic Map